Invitrogen
Manufacturer Code:PA511926
Catalog # PIPA511926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human SNAI1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein sna; Protein snail homolog 1; snail 1 homolog; snail 1 zinc finger protein; snail 1, zinc finger protein; snail family zinc finger 1; snail homolog 1; Snail protein homolog Sna protein; Zinc finger protein SNAI1
Gene Aliases: AI194338; dJ710H13.1; SLUGH2; SNA; Sna1; SNAH; SNAI1; SNAIL; SNAIL1
UniProt ID: (Human) O95863, (Mouse) Q02085
Entrez Gene ID: (Human) 6615, (Mouse) 20613
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.