Proteintech
Manufacturer Code:175941AP150UL
Catalog # 50-173-1673
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SMYD4 Fusion Protein Ag11552 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1936 SMYD4 ZMYND21; SET and MYND domain-containing protein 4
Gene Aliases: G430029E23Rik; KIAA1936; SMYD4; ZMYND21
UniProt ID: (Human) Q8IYR2, (Mouse) Q8BTK5
Entrez Gene ID: (Human) 114826, (Mouse) 319822, (Rat) 287525
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.