Invitrogen
Manufacturer Code:PA5115938
Catalog # PIPA5115938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SMYD2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110020E07Rik 4930402C15 Histone methyltransferase SMYD2 HSKM-B KMT3C Lysine N-methyltransferase 3C N-lysine methyltransferase SMYD2 SET and MYND domain containing 2 SET and MYND domain-containing protein 2 SMYD2 zinc finger MYND domain containing 14 ZMYND14; Histone methyltransferase SMYD2; HSKM-B; Lysine N-methyltransferase 3C; N-lysine methyltransferase SMYD2; SET and MYND domain-containing protein 2; zinc finger, MYND domain containing 14
Gene Aliases: 1110020E07Rik; 4930402C15; HSKM-B; KMT3C; SMYD2; ZMYND14
UniProt ID: (Human) Q9NRG4, (Rat) Q7M6Z3, (Mouse) Q8R5A0
Entrez Gene ID: (Human) 56950, (Rat) 289372, (Mouse) 226830
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.