Novus Biologicals
Manufacturer Code:NBP299700AF350
Catalog # NB089053
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant SMYD2 (NP_064582.2 Met 1-His 433). |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.43 Histone methyltransferase SMYD2 HSKM-Bzinc finger MYND domain containing 14 KMT3CEC 2.1.1.- Lysine N-methyltransferase 3C MGC119305 SET and MYND domain containing 2 SET and MYND domain-containing protein 2 ZMYND14; Histone methyltransferase SMYD2; HSKM-B; Lysine N-methyltransferase 3C; N-lysine methyltransferase SMYD2; SET and MYND domain-containing protein 2; zinc finger, MYND domain containing 14
Gene Aliases: HSKM-B; KMT3C; SMYD2; ZMYND14
UniProt ID: (Human) Q9NRG4
Entrez Gene ID: (Human) 56950
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.