R&D Systems
Manufacturer Code:MAB9507SP
Catalog # MAB9507SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 530131 |
Immunogen | <em>E. coli</em>-derived recombinant human SMURF1 Met496-Glu757 Accession # Q9HCE7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: E3 ubiquitin ligase SMURF1; E3 ubiquitin ligase SMURF1 EC 6.3.2 EC 6.3.2.- hSMURF1 KIAA1625 KIAA1625E3 ubiquitin-protein ligase SMURF1 SMAD specific E3 ubiquitin protein ligase 1 SMAD ubiquitination regulatory factor 1 Smad-specific E3 ubiquitin ligase 1 SMAD-specific E3 ubiquitin-protein ligase 1; E3 ubiquitin-protein ligase SMURF1; HECT-type E3 ubiquitin transferase SMURF1; hSMURF1; SMAD ubiquitination regulatory factor 1; Smad-specific E3 ubiquitin ligase 1; SMAD-specific E3 ubiquitin-protein ligase 1
Gene Aliases: KIAA1625; SMURF1
UniProt ID: (Human) Q9HCE7
Entrez Gene ID: (Human) 57154
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.