Novus Biologicals
Manufacturer Code:NBP152981
Catalog # NBP152981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SMUG1(single-strand-selective monofunctional uracil-DNA glycosylase 1) The peptide sequence was selected from the middle region of SMUG1. Peptide sequence IVGPVLTPPQEHPKRPVLGLECPQSEVSGARFWGFFRNLCGQPEVFFH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.2.2 EC 3.2.2.- FDG HMUDG MGC104370 single-strand selective monofunctional uracil DNA glycosylase single-strand-selective monofunctional uracil-DNA glycosylase 1 UNG3; Single-strand selective monofunctional uracil DNA glycosylase
Gene Aliases: FDG; HMUDG; SMUG1; UNG3
UniProt ID: (Human) Q53HV7
Entrez Gene ID: (Human) 23583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.