OriGene
Manufacturer Code:TA890024
Catalog # 50-168-4737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human SMUG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FDG HMUDG UNG3; Single-strand selective monofunctional uracil DNA glycosylase; Single-strand selective monofunctional uracil-DNA glycosylase
Gene Aliases: 1200013B09Rik; A930006H09Rik; C85220; FDG; HMUDG; SMUG1; UNG3
UniProt ID: (Human) Q53HV7, (Mouse) Q6P5C5, (Rat) Q811Q1
Entrez Gene ID: (Human) 23583, (Mouse) 71726, (Rat) 315344
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.