Invitrogen
Manufacturer Code:PA571762
Catalog # PIPA571762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide selected from the N-terminal region of Drosophila SUMO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG4494-PA; CG4494-PA LD07775p Smt3 Ubiquitin-like protein SMT3; CG4494-PB; LD07775p; lethal (2) SH0182; small Ubiquitin-like modifier; smt3-PA; smt3-PB; SUMO; SUMO protein
Gene Aliases: anon-EST:Posey240; CG4494; Dm0342; Dmel\CG4494; Dmel_CG4494; DmSmt3; DmSUMO-1; dSmt3; l(2)04493; l(2)SH0182; l(2)SH2 0182; SMT3; SUMO
Entrez Gene ID: (Fruit fly) 33981
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.