Abnova
Manufacturer Code:PAB10385
Catalog # 89-114-825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Drosophila SMSr. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SMSr.
For Research Use Only
Protein Aliases: Ceramide phosphoethanolamine synthase; CG32380; CG32380-PA; CG8572; CG8576; CPE synthase; Css3beta; GH24627p; SMSr-PA; Sphingomyelin synthase-related 1
Gene Aliases: CG32380; CG8572; CG8576; Css3beta; Dmel\CG32380; Dmel_CG32380; dmSMSr; dSMSr; SMSr
UniProt ID: (Fruit fly) Q9VS60
Entrez Gene ID: (Fruit fly) 38823
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.