OriGene
Manufacturer Code:TA700122
Catalog # 50-167-7658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5C12 |
Immunogen | Full length human recombit protein of human SMS produced in HEK293T cell. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MRSR SPMSY SRS SpS; Spermidine aminopropyltransferase; Spermine synthase; SPMSY
Gene Aliases: MRSR; SMS; SPMSY; SpS; SRS
UniProt ID: (Human) P52788
Entrez Gene ID: (Human) 6611
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.