Invitrogen
Manufacturer Code:PA540798
Catalog # PIPA540798
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SMPDL3B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid sphingomyelinase-like phosphodiesterase 3b; ASM-like phosphodiesterase 3b; sphingomyelin phosphodiesterase acid-like 3B; sphingomyelin phosphodiesterase, acid-like 3B
Gene Aliases: 1110054A24Rik; ASML3B; ASMLPD; AU045240; im:7137990; si:ch1073-55d10.1; SMPDL3B
UniProt ID: (Human) Q92485, (Mouse) P58242
Entrez Gene ID: (Dog) 100855784, (Human) 27293, (Horse) 100070816, (Bovine) 518699, (Rabbit) 100342054, (Rat) 362619, (Guinea pig) 100735193, (Mouse) 100340, (Zebrafish) 564392
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.