Abnova
Manufacturer Code:PAB18796
Catalog # 89-120-353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 15 amino acids at internal region of human SMPDL3B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SMPDL3B.
For Research Use Only
Protein Aliases: Acid sphingomyelinase-like phosphodiesterase 3b; ASM-like phosphodiesterase 3b; ASML3B; sphingomyelin phosphodiesterase acid-like 3B; sphingomyelin phosphodiesterase, acid-like 3B
Gene Aliases: ASML3B; ASMLPD; SMPDL3B
UniProt ID: (Human) Q92485
Entrez Gene ID: (Human) 27293
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.