Invitrogen
Manufacturer Code:PA569826
Catalog # PIPA569826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse SMPDL3A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610010C24Rik; Acid sphingomyelinase-like phosphodiesterase 3a; ASM-like phosphodiesterase 3a; ASM3A ASML3a yR36GH4.1 0610010C24Rik ASM-like phosphodiesterase 3a acid sphingomyelinase-like phosphodiesterase 3a; sphingomyelin phosphodiesterase acid-like 3A; sphingomyelin phosphodiesterase, acid-like 3A
Gene Aliases: 0610010C24Rik; AI529588; ASM3A; ASML3; ASML3A; si:ch211-199i15.3; SMPDL3A; yR36GH4.1; zgc:113352
UniProt ID: (Human) Q92484, (Bovine) Q3ZC91, (Rat) Q641Z7, (Mouse) P70158
Entrez Gene ID: (Human) 10924, (Rabbit) 100358362, (Horse) 100072974, (Bovine) 505300, (Dog) 476279, (Rat) 294422, (Mouse) 57319, (Guinea pig) 100730319, (Zebrafish) 541525
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.