Invitrogen
Manufacturer Code:PA561204
Catalog # PIPA561204
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SMPD4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NET13 NSMASE-3 NSMASE3 SKNY protein neutral sphingomyelinase 3 neutral sphingomyelinase III sphingomyelin phosphodiesterase 4; Neutral sphingomyelinase 3; Neutral sphingomyelinase III; neutral sphingomyelinase-3; nSMase-3; SKNY protein; Sphingomyelin phosphodiesterase 4; sphingomyelin phosphodiesterase 4, neutral membrane (neutral sphingomyelinase-3)
Gene Aliases: KIAA1418; NET13; NSMASE-3; NSMASE3; SKNY; SMPD4
UniProt ID: (Human) Q9NXE4
Entrez Gene ID: (Human) 55627
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.