Novus Biologicals
Manufacturer Code:NBP2857830.1ML
Catalog # NB033712
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Rat SMPD4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1418 NET13 Neutral Sphingomyelinase 3 Neutral Sphingomyelinase III NSMASE3 NSMASE-3 SKNY Protein Sphingomyelin Phosphodiesterase 4 Sphingomyelin Phosphodiesterase 4 Neutral Membrane (Neutral Sphingomyelinase-3); Neutral sphingomyelinase 3; Neutral sphingomyelinase III; neutral sphingomyelinase-3; nSMase-3; SKNY protein; Sphingomyelin phosphodiesterase 4; sphingomyelin phosphodiesterase 4, neutral membrane (neutral sphingomyelinase-3)
Gene Aliases: KIAA1418; NET13; NSMASE-3; NSMASE3; SKNY; SMPD4
UniProt ID: (Human) Q9NXE4
Entrez Gene ID: (Human) 55627
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.