Novus Biologicals
Manufacturer Code:NBP222365100UL
Catalog # NBP22236510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH-conjugated synthetic peptide mapping to a fragment of residues within amino acids 391-419 in the C-terminal region of human SMPD1 UniProt Accession No.P17405. Species sequence homology: bovine |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: Acid sphingomyelinase; Acid sphingomyelinase ASMASE ASMsphingomyelin phosphodiesterase EC 3.1.4.12 NPD sphingomyelin phosphodiesterase 1 acid lysosomal; aSMase; Sphingomyelin phosphodiesterase; sphingomyelin phosphodiesterase 1, acid lysosomal
Gene Aliases: ASM; ASMASE; NPD; SMPD1
UniProt ID: (Human) P17405
Entrez Gene ID: (Human) 6609
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.