Invitrogen
Manufacturer Code:PA572395
Catalog # PIPA572395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | region 85-116 aa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bent body; frizzled family member 11; Protein Gx; seven transmembrane helix receptor; SMO; SMO smoothened homolog (Drosophila) HGNC:11119 Gx SMOH seven transmembrane helix receptor smoothened; Smoothened homolog; smoothened, frizzled family receptor; smoothened, seven transmembrane spanning receptor
Gene Aliases: bnb; E130215L21Rik; FZD11; Gx; SMO; SMOH; smoothened
UniProt ID: (Human) Q99835, (Mouse) P56726, (Rat) P97698
Entrez Gene ID: (Human) 6608, (Mouse) 319757, (Rat) 25273
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.