Invitrogen
Manufacturer Code:PA534217
Catalog # PIPA534217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 15 amino acid peptide from N-terminal extracellular domain of human SMO. Percent identity with other species by BLAST analysis: Human Gorilla Gibbon Monkey Marmoset Mouse Rat Bovine Dog Bat Hamster Elephant Panda Horse Rabbit Pig Platypus (100&percnt) Opossum (93&percnt) Turkey Chicken Xenopus (87&percnt) Zebrafish Seq squirt (80&percnt). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: frizzled family member 11; Protein Gx; seven transmembrane helix receptor; SMO; SMO Frizzled family member 11 FZD11 Gx Protein Gx SMOH Smoothened Smoothened homolog; Smoothened homolog; smoothened, frizzled family receptor; smoothened, seven transmembrane spanning receptor
Gene Aliases: FZD11; Gx; SMO; SMOH
UniProt ID: (Human) Q99835
Entrez Gene ID: (Human) 6608
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.