Invitrogen
Manufacturer Code:PA5104390
Catalog # PIPA5104390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SMO corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bent body; bent body bnb E130215L21Rik frizzled family member 11 FZD11 Gx HGNC:11119 protein Gx Protein SMOH seven transmembrane helix receptor SMO SMOH smoothened Smoothened homolog smoothened homolog (Drosophila) smoothened frizzled class receptor smoothened frizzled family receptor smoothened seven transmembrane spanning receptor; frizzled family member 11; Protein Gx; seven transmembrane helix receptor; SMO; Smoothened homolog; smoothened, frizzled family receptor; smoothened, seven transmembrane spanning receptor
Gene Aliases: bnb; E130215L21Rik; FZD11; Gx; SMO; SMOH; smoothened
UniProt ID: (Human) Q99835, (Mouse) P56726, (Rat) P97698
Entrez Gene ID: (Human) 6608, (Mouse) 319757, (Rat) 25273
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.