Abnova
Manufacturer Code:H00006608A01
Catalog # 89-105-472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 56 a.a. &bsim 155 a.a) partial recombinant protein with GST tag.,SMO (NP_005622 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SMO.
For Research Use Only
Protein Aliases: frizzled family member 11; Gx; Protein Gx; seven transmembrane helix receptor; SMO; SMOH; Smoothened homolog; smoothened, frizzled family receptor; smoothened, seven transmembrane spanning receptor
Gene Aliases: FZD11; Gx; SMO; SMOH
UniProt ID: (Human) Q99835
Entrez Gene ID: (Human) 6608
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.