Invitrogen
Manufacturer Code:PA531840
Catalog # PIPA531840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 719 and 1003 of Human SMG7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: breast cancer-associated antigen SGA-56M; C1orf16 EST1C SGA56M EST1 telomerase component homolog C EST1-like protein C breast cancer-associated antigen SGA-56M ever shorter telomeres 1C protein SMG7; EST1 telomerase component homolog C; EST1-like protein C; ever shorter telomeres 1C; hSMG-7; Protein SMG7; SMG-7 homolog; smg-7 homolog, nonsense mediated mRNA decay factor; SMG7 nonsense mediated mRNA decay factor
Gene Aliases: 9430023P16Rik; C1orf16; EST1C; KIAA0250; mKIAA0250; SGA56M; SMG7
UniProt ID: (Human) Q92540, (Mouse) Q5RJH6
Entrez Gene ID: (Human) 9887, (Mouse) 226517
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.