Novus Biologicals
Manufacturer Code:NBP220423
Catalog # NBP220423
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human SMG7. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: breast cancer-associated antigen SGA-56M; breast cancer-associated antigen SGA-56M C1orf16 chromosome 1 open reading frame 16 EST1 telomerase component homolog C EST1CKIAA0250SGA56M EST1-like protein C ever shorter telomeres 1C FLJ23717 hSMG-7 protein SMG7 SMG-7 SMG-7 homolog Smg-7 homolog nonsense mediated mRNA decay factor (C. elegans); EST1 telomerase component homolog C; EST1-like protein C; ever shorter telomeres 1C; hSMG-7; Protein SMG7; SMG-7 homolog; smg-7 homolog, nonsense mediated mRNA decay factor; SMG7 nonsense mediated mRNA decay factor
Gene Aliases: C1orf16; EST1C; KIAA0250; SGA56M; SMG7
UniProt ID: (Human) Q92540
Entrez Gene ID: (Human) 9887
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.