Abnova
Manufacturer Code:PAB17426
Catalog # 89-119-084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human SMG7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SMG7.
For Research Use Only
Protein Aliases: breast cancer-associated antigen SGA-56M; C1orf16; EST1 telomerase component homolog C; EST1-like protein C; EST1C; ever shorter telomeres 1C; FLJ23717; hSMG-7; KIAA0250; Protein SMG7; SGA56M; SMG-7; SMG-7 homolog; smg-7 homolog, nonsense mediated mRNA decay factor; SMG7 nonsense mediated mRNA decay factor
Gene Aliases: C1orf16; EST1C; KIAA0250; SGA56M; SMG7
UniProt ID: (Human) Q92540
Entrez Gene ID: (Human) 9887
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.