Invitrogen
Manufacturer Code:PA551545
Catalog # PIPA551545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SMEK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: protein phosphatase 4, regulatory subunit 3B; Serine/threonine-protein phosphatase 4 regulatory subunit 3B; Serine/threonine-protein phosphatase 4 regulatory subunit 3B SMEK homolog 2 SMEK2 KIAA1387 PP4R3B PPP4R3B; SMEK homolog 2; SMEK homolog 2, suppressor of mek1
Gene Aliases: AW011752; AW557776; FLFL2; KIAA1387; mKIAA1387; PP4R3B; PPP4R3B; PSY2; RGD1309450; SMEK2; smk1
UniProt ID: (Human) Q5MIZ7, (Mouse) Q922R5
Entrez Gene ID: (Human) 57223, (Mouse) 104570, (Rat) 360993
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.