Invitrogen
Manufacturer Code:PA556934
Catalog # PIPA556934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Human SMDT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Store at &minus20°C long term. Avoid freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Essential MCU regulator, mitochondrial; single-pass membrane protein with aspartate-rich tail 1; Single-pass membrane protein with aspartate-rich tail 1, mitochondrial; UPF0466 protein C22orf32, mitochondrial
Gene Aliases: C22orf32; DDDD; dJ186O1.1; EMRE; SMDT1
UniProt ID: (Human) Q9H4I9
Entrez Gene ID: (Human) 91689
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.