Novus Biologicals
Manufacturer Code:NBP255009
Catalog # NBP255009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LGLMAKDKTLRKKDKEGPECPPTVVVKEELGGDVKVESTSPKTFLESKEELSHSPEPCTKMTMRLRRNHSNAQFIESYVCRMCSRG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(4) demethylase 5C; DXS1272EMRXJ EC 1.14.11 EC 1.14.11.- Histone demethylase JARID1C JARID1Clysine-specific demethylase 5C jumonji AT rich interactive domain 1C Jumonji AT rich interactive domain 1C (RBP2-like) Jumonji/ARID domain-containing protein 1C lysine (K)-specific demethylase 5C MRXSJ Protein SmcX Protein Xe169 Smcx homolog X chromosome SMCXselected cDNA on X Smcy homolog X-linked Smcy homolog X-linked (mouse) XE169JmjC domain-containing protein SMCX; Histone demethylase JARID1C; JmjC domain-containing protein SMCX; Jumonji, AT rich interactive domain 1C (RBP2-like); Jumonji/ARID domain-containing protein 1C; lysine (K)-specific demethylase 5C; Lysine-specific demethylase 5C; Protein SmcX; Protein Xe169; Smcx homolog, X chromosome; Smcy homolog, X-linked
Gene Aliases: DXS1272E; JARID1C; KDM5C; MRX13; MRXJ; MRXSCJ; MRXSJ; SMCX; XE169
UniProt ID: (Human) P41229
Entrez Gene ID: (Human) 8242
Molecular Function: transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.