Novus Biologicals
Manufacturer Code:NBP274239AF350
Catalog # NP274239A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6G9 |
Immunogen | Full length recombinant protein of human SMCP (NP_109588) produced in E.coli. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: HSMCSGEN1 MCSMGC26305 MCSPMGC26519 mitochondrial capsule selenoprotein sperm mitochondria-associated cysteine-rich protein sperm mitochondrial-associated cysteine-rich protein; mitochondrial capsule selenoprotein; sperm mitochondria-associated cysteine-rich protein; Sperm mitochondrial-associated cysteine-rich protein; testicular tissue protein Li 119; testicular tissue protein Li 161
Gene Aliases: HSMCSGEN1; MCS; MCSP; SMCP
UniProt ID: (Human) P49901
Entrez Gene ID: (Human) 4184
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.