Abnova
Manufacturer Code:H00004184M04A
Catalog # 89-104-900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F4 |
Immunogen | SMCP (AAH145931 a.a. &bsim 116 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant SMCP.
For Research Use Only
Protein Aliases: HSMCSGEN1; MCS; MCSP; MGC26305; MGC26519; mitochondrial capsule selenoprotein; sperm mitochondria-associated cysteine-rich protein; Sperm mitochondrial-associated cysteine-rich protein; testicular tissue protein Li 119; testicular tissue protein Li 161
Gene Aliases: HSMCSGEN1; MCS; MCSP; SMCP
UniProt ID: (Human) P49901
Entrez Gene ID: (Human) 4184
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.