Novus Biologicals
Manufacturer Code:NB100470
Catalog # NB100470
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 1050 and the C-terminus (residue 1091) of human Structural Maintenance of Chromosomes 6 using the numbering given in TrEMBL entry Q96SB8 (GeneID 79677). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ22116 hSMC6 SMC protein 6 SMC-6 SMC6 structural maintenance of chromosomes 6-like 1 SMC6 structural maintenance of chromosomes 6-like 1 (yeast) SMC6L1FLJ35534 structural maintenance of chromosomes 6 structural maintenance of chromosomes protein 6; SMC protein 6; SMC6 structural maintenance of chromosomes 6-like 1; Structural maintenance of chromosomes protein 6
Gene Aliases: hSMC6; SMC-6; SMC6; SMC6L1
UniProt ID: (Human) Q96SB8
Entrez Gene ID: (Human) 79677
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.