Invitrogen
Manufacturer Code:PA545522
Catalog # PIPA545522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SMC6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mSMC6; SMC protein 6; SMC6 structural maintenance of chromosomes 6-like 1; Structural maintenance of chromosomes protein 6; Structural maintenance of chromosomes protein 6-like protein
Gene Aliases: 2810489L22Rik; 3830418C19Rik; AA990493; AU018782; AW742439; hSMC6; Kiaa4103; mKIAA4103; SMC-6; SMC6; SMC6L1
UniProt ID: (Human) Q96SB8, (Mouse) Q924W5
Entrez Gene ID: (Dog) 475675, (Horse) 100072064, (Human) 79677, (Rabbit) 100354764, (Rat) 313961, (Mouse) 67241, (Guinea pig) 100721259
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.