Invitrogen
Manufacturer Code:PA530820
Catalog # PIPA530820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 526 and 776 of Human SMC6L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SMC protein 6; SMC-6 SMC6L1 hSMC6 SMC protein 6 SMC6 structural maintece of chromosomes 6-like 1 structural maintece of chromosomes protein 6; SMC6 structural maintenance of chromosomes 6-like 1; Structural maintenance of chromosomes protein 6
Gene Aliases: hSMC6; SMC-6; SMC6; SMC6L1
UniProt ID: (Human) Q96SB8
Entrez Gene ID: (Human) 79677
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.