Invitrogen
Manufacturer Code:PA530460
Catalog # PIPA530460
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 257 and 493 of Human SMC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSMC5; MLZ-453; Protein expressed in male leptotene and zygotene spermatocytes 453; SMC protein 5; SMC-5; SMC5 structural maintenance of chromosomes 5-like 1; SMC5L1 SMC protein 5 SMC-5 SMC5 structural maintece of chromosomes 5-like 1 hSMC5 structural maintece of chromosomes protein 5; Structural maintenance of chromosomes protein 5
Gene Aliases: KIAA0594; mKIAA0594; MLZ-453; mSMC5; SMC-5; SMC5; SMC5L1
UniProt ID: (Human) A6NM81, (Mouse) Q8CG46
Entrez Gene ID: (Human) 23137, (Mouse) 226026
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.