Novus Biologicals
Manufacturer Code:NB100469
Catalog # NB100469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 1050 and the C-terminus (residue 1101) of human Structural Maintenance of Chromosomes 5 using the numbering given in TrEMBL entry Q8IY18 (GeneID 23137). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSMC5; hSMC5 KIAA0594SMC5 structural maintenance of chromosomes 5-like 1 (yeast) SMC protein 5 SMC-5 SMC5L1SMC5 structural maintenance of chromosomes 5-like 1 structural maintenance of chromosomes 5 structural maintenance of chromosomes protein 5; SMC protein 5; SMC-5; SMC5 structural maintenance of chromosomes 5-like 1; Structural maintenance of chromosomes protein 5
Gene Aliases: KIAA0594; SMC5; SMC5L1
UniProt ID: (Human) A6NM81
Entrez Gene ID: (Human) 23137
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.