Abnova
Manufacturer Code:H00023137K
Catalog # 89-201-322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SMC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SMC5 peptide using ARM Technology.
For Research Use Only
Protein Aliases: hSMC5; KIAA0594 SMC5L1; SMC protein 5; SMC-5; SMC5 structural maintenance of chromosomes 5-like 1; Structural maintenance of chromosomes protein 5
Gene Aliases: KIAA0594; SMC5; SMC5L1
UniProt ID: (Human) A6NM81
Entrez Gene ID: (Human) 23137
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.