Novus Biologicals
Manufacturer Code:NBP297937IR
Catalog # NB049656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SMC4 fragment. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAP-C CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast) Chromosome-associated polypeptide C hCAP-CSMC4 (structural maintenance of chromosomes 4 yeast)-like 1 SMC protein 4 SMC-4 SMC4 structural maintenance of chromosomes 4-like 1 SMC4L1 structural maintenance of chromosomes 4 structural maintenance of chromosomes protein 4 XCAP-C homolog; Chromosome-associated polypeptide C; hCAP-C; SMC protein 4; SMC4 structural maintenance of chromosomes 4-like 1; Structural maintenance of chromosomes protein 4; XCAP-C homolog
Gene Aliases: CAP-C; CAPC; SMC-4; SMC4; SMC4L1
UniProt ID: (Human) Q9NTJ3
Entrez Gene ID: (Human) 10051
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.