Invitrogen
Manufacturer Code:PA141402
Catalog # 01-675-531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 281-297 of human CAP-C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAPC SMC4L1 hCAP-C; Chromosome-associated polypeptide C; hCAP-C; SMC protein 4; SMC4 structural maintenance of chromosomes 4-like 1; Structural maintenance of chromosomes protein 4; XCAP-C homolog
Gene Aliases: CAP-C; CAPC; SMC-4; SMC4; SMC4L1
UniProt ID: (Human) Q9NTJ3
Entrez Gene ID: (Human) 10051
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.