Novus Biologicals
Manufacturer Code:NBP267153
Catalog # NBP267153
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM10-75 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Bamacan; Bamacan BAMSMC protein 3 Basement membrane-associated chondroitin proteoglycan BMH Chondroitin sulfate proteoglycan 6 chondroitin sulfate proteoglycan 6 (bamacan) Chromosome-associated polypeptide CSPG6structural maintenance of chromosomes protein 3 hCAP HCAPbamacan SMC-3 SMC3L1CDLS3 structural maintenance of chromosomes 3; Basement membrane-associated chondroitin proteoglycan; Chondroitin sulfate proteoglycan 6; chondroitin sulfate proteoglycan 6 (bamacan); Chromosome-associated polypeptide; hCAP; SMC protein 3; Structural maintenance of chromosomes protein 3
Gene Aliases: BAM; BMH; CDLS3; CSPG6; HCAP; SMC3; SMC3L1
UniProt ID: (Human) Q9UQE7
Entrez Gene ID: (Human) 9126
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.