Invitrogen
Manufacturer Code:PA529131
Catalog # PIPA529131
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 909 and 1199 of Human SMC3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAM BMH CDLS3 CSPG6 HCAP SMC3L1 SMC protein 3 SMC-3 basement membrane-associated chondroitin proteoglycan chondroitin sulfate proteoglycan 6 (bamacan) chromosome-associated polypeptide structural maintece of chromosomes protein 3; Bamacan; basement membrane chondroitin sulfate proteoglycan; Basement membrane-associated chondroitin proteoglycan; Chondroitin sulfate proteoglycan 6; chondroitin sulfate proteoglycan 6 (bamacan); Chromosome segregation protein SmcD; Chromosome-associated polypeptide; hCAP; Mad member-interacting protein 1; SMC protein 3; structural maintenace of chromosomes 3; Structural maintenance of chromosomes protein 3
Gene Aliases: BAM; Bamacan; BMH; CDLS3; CSPG6; HCAP; Mmip1; SMC-3; SMC3; SMC3L1; SmcD
UniProt ID: (Human) Q9UQE7, (Mouse) Q9CW03
Entrez Gene ID: (Human) 9126, (Mouse) 13006, (Rat) 29486
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.