OriGene
Manufacturer Code:TA809793
Catalog # 50-168-3026
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5A1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 889-1016 of human SMC1A produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CDLS2 DXS423E SB1.8 SMC1 SMC1L1 SMC1alpha SMCB; Chromosome segregation protein SmcB; Sb1.8; segregation of mitotic chromosomes 1; segregation of mitotic chromosomes b; SMC (segregation of mitotic chromosomes 1)-like 1; SMC protein 1A; SMC-1-alpha; SMC1 (structural maintenance of chromosomes 1, yeast)-like 1; Structural maintenance of chromosomes protein 1A
Gene Aliases: 5830426I24Rik; CDLS2; DXS423E; KIAA0178; mKIAA0178; SB1.8; SMC-1A; SMC1; SMC1A; SMC1alpha; SMC1L1; SMCB
UniProt ID: (Human) Q14683, (Mouse) Q9CU62
Entrez Gene ID: (Human) 8243, (Mouse) 24061
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.