Abnova
Manufacturer Code:MAB2014
Catalog # 89-110-758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | C2M |
Immunogen | Recombinant protein corresponding to amino acids 402-894 of human SMC1A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant SMC1A.
For Research Use Only
Protein Aliases: CDLS2; DKFZp686L19178; DXS423E; KIAA0178; MGC138332; SB1.8; segregation of mitotic chromosomes 1; SMC protein 1A; SMC-1-alpha; SMC1; SMC1 (structural maintenance of chromosomes 1, yeast)-like 1; SMC1alpha; SMC1L1; SMCB; Structural maintenance of chromosomes protein 1A
Gene Aliases: CDLS2; DXS423E; KIAA0178; SB1.8; SMC1; SMC1A; SMC1alpha; SMC1L1; SMCB
UniProt ID: (Human) Q14683
Entrez Gene ID: (Human) 8243
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.