Novus Biologicals
Manufacturer Code:NBP276412
Catalog # NBP276412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | BL-205-2G8 |
Immunogen | Immunogen sequence between amino acids 1200 and 1233 (C-Term) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2-8°C/ 1 year from date of receipt |
Rabbit Monoclonal Antibody
Protein Aliases: CDLS2 DXS423ESMC1 structural maintenance of chromosomes 1-like 1 (yeast) KIAA0178DKFZp686L19178 Sb1.8 SB1.8MGC138332 segregation of mitotic chromosomes 1 SMC protein 1A SMC1 (structural maintenance of chromosomes 1 yeast)-like 1 SMC1 structural maintenance of chromosomes 1-like 1 SMC-1A SMC-1-alpha SMC1L1 SMC1SMC1alpha SMCB structural maintenance of chromosomes 1A structural maintenance of chromosomes protein 1A; Sb1.8; segregation of mitotic chromosomes 1; SMC protein 1A; SMC-1-alpha; SMC1 (structural maintenance of chromosomes 1, yeast)-like 1; Structural maintenance of chromosomes protein 1A
Gene Aliases: CDLS2; DXS423E; KIAA0178; SB1.8; SMC1; SMC1A; SMC1alpha; SMC1L1; SMCB
UniProt ID: (Human) Q14683
Entrez Gene ID: (Human) 8243
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.