Invitrogen
Manufacturer Code:PA541441
Catalog # PIPA541441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SMARCD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60 kDa BRG-1/Brm-associated factor subunit B; BAF60B; BRG1-associated factor 60B; chromatin remodeling complex BAF60B subunit; mammalian chromatin remodeling complex BRG1-associated factor 60B; SWI/SNF complex 60 kDa subunit B; SWI/SNF related matrix associated actin dependent regulator of Chromatin subfamily d member 2 BAF60B CRACD2 PRO2451 Rsc6p; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 2; Swp73-like protein
Gene Aliases: AW322457; BAF60B; CRACD2; PRO2451; Rsc6p; SMARCD2
UniProt ID: (Bovine) E1BJD1, (Human) Q92925, (Mouse) Q99JR8, (Rat) O54772
Entrez Gene ID: (Horse) 100053934, (Dog) 480477, (Bovine) 789613, (Rabbit) 100350037, (Human) 6603, (Mouse) 83796, (Rat) 83833, (Zebrafish) 564317
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.