Novus Biologicals
Manufacturer Code:NBP298471AF594
Catalog # NB054072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SMARCD1 fragment. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60 kDa BRG-1/Brm-associated factor subunit A; BAF60A; BAF60Amammalian chromatin remodeling complex BRG1-associated factor 60A BRG1-associated factor 60A chromatin remodeling complex BAF60A subunit CRACD1 Rsc6p subfamily d member 1 SWI/SNF complex 60 kDa subunit SWI/SNF complex 60 kDa subunit A SWI/SNF related matrix associated actin dependent regulator of chromatin SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 160 kDa BRG-1/Brm-associated factor subunit A Swp73-like protein; BRG1-associated factor 60A; chromatin remodeling complex BAF60A subunit; mammalian chromatin remodeling complex BRG1-associated factor 60A; SMARCD1; SWI/SNF complex 60 kDa subunit; SWI/SNF complex 60 kDa subunit A; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 1; Swp73-like protein
Gene Aliases: BAF60A; CRACD1; Rsc6p; SMARCD1
UniProt ID: (Human) Q9Y635
Entrez Gene ID: (Human) 6602
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.