Invitrogen
Manufacturer Code:PA581033
Catalog # PIPA581033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the center region of the Human SMARCC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAF155; BAF155 CRACC1 Rsc8 SRG3 SWI3 BRG1-associated factor 155 SWI/SNF complex 155 kDa subunit SWI/SNF complex subunit SMARCC1 SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 1 chromatin remodeling complex BAF155 subunit mammalian chromatin remodeling complex BRG1-associated factor 155; BRG1-associated factor 155; chromatin remodeling complex BAF155 subunit; mammalian chromatin remodeling complex BRG1-associated factor 155; SWI/SNF complex 155 kDa subunit; SWI/SNF complex subunit SMARCC1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 1
Gene Aliases: BAF155; CRACC1; Rsc8; SMARCC1; SRG3; SWI3
UniProt ID: (Human) Q92922
Entrez Gene ID: (Human) 6599
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.