Invitrogen
Manufacturer Code:PA5115941
Catalog # PIPA5115941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SNF5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU020204 BAF47 BRG1-associated factor 47 CSS3 hSNF5 hSNFS INI1 Integrase interactor 1 protein malignant rhabdoid tumor suppressor MRD15 mSNF5 PPP1R144 protein phosphatase 1 regulatory subunit 144 RDT RTPS1 Sfh1p SMARCB1 SNF5 SNF5 homolog SNF5/INI1 SNF5L1 Snr1 sucrose nonfermenting yeast homolog-like 1 SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily b member 1 SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1 SWI/SNF-related matrix-associated protein SWI10 SWNTS1 TYE4; BAF47; BRG1-associated factor 47; hSNF5; Integrase interactor 1 protein; malignant rhabdoid tumor suppressor; mSNF5; protein phosphatase 1, regulatory subunit 144; SNF5 homolog; sucrose nonfermenting, yeast, homolog-like 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1; SWI/SNF-related matrix-associated protein
Gene Aliases: AU020204; BAF47; CSS3; hSNFS; INI1; MRD15; PPP1R144; RDT; RTPS1; Sfh1p; SMARCB1; SNF5; SNF5/INI1; SNF5L1; Snr1; SWNTS1
UniProt ID: (Human) Q76N08, (Mouse) Q9Z0H3
Entrez Gene ID: (Human) 6598, (Mouse) 20587, (Rat) 361825
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.