Invitrogen
Manufacturer Code:PA540914
Catalog # PIPA540914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SMARCAD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase 1; ATP-dependent helicase SMARCAD1; enhancer trap locus 1; Enhancer trap locus homolog 1; Etl-1; hHEL1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A containing DEAD/H box 1
Gene Aliases: ADERM; AV081750; AW226546; D6Pas1; ETL1; HEL1; KIAA1122; mKIAA1122; SMARCAD1
UniProt ID: (Human) Q9H4L7, (Rat) D3Z9Z9, (Mouse) Q04692
Entrez Gene ID: (Rabbit) 100354709, (Bovine) 530506, (Horse) 100053412, (Pig) 100515330, (Human) 56916, (Dog) 100856284, (Rat) 312398, (Mouse) 13990
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.