Novus Biologicals
Manufacturer Code:NB10079834
Catalog # NB10079834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogenfor this product falls between residues 1 and 50 of human SWI/SNFrelated matrix-associated actin-dependent regulator of chromatin subfamily A containing DEAD/H box 1 (enhancer trap locus 1) using the numbering given in entry NP_064544.1 (GeneID 56916). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase 1; ATP-dependent helicase 1 DKFZP762K2015 EC 3.6.1 EC 3.6.4.12 HEL1 hHEL1 KIAA1122ETL1DKFZp762K2015 subfamily a containing DEAD/H box 1 SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A containing DEAD/H box 1 SWI/SNF-related matrix-associated actin-dependent regulator of chromatin; hHEL1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A containing DEAD/H box 1
Gene Aliases: ADERM; ETL1; HEL1; KIAA1122; SMARCAD1
UniProt ID: (Human) Q9H4L7
Entrez Gene ID: (Human) 56916
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.