Invitrogen
Manufacturer Code:MA535378
Catalog # PIMA535378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0795 |
Immunogen | A synthesized peptide derived from human SNF2H |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 4933427E24Rik BOS_16311 D030040M08Rik D330027N15Rik hISWI hSNF2H ISWI MommeD4 mSnf2h Smarca5 SNF2H Sucrose nonfermenting protein 2 homolog SWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5 SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 5 WCRF135; hSNF2H; mSnf2h; Sucrose nonfermenting protein 2 homolog; sucrose nonfermenting-like 5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 5
Gene Aliases: 4933427E24Rik; D030040M08Rik; D330027N15Rik; hISWI; hSNF2H; ISWI; MommeD4; SMARCA5; SNF2H; WCRF135
UniProt ID: (Human) O60264, (Mouse) Q91ZW3
Entrez Gene ID: (Human) 8467, (Rat) 307766, (Mouse) 93762
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.