Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated. View details.
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | 0.3 µg |
Western Blot (WB) | 1:1,000 |
Immunocytochemistry (ICC/IF) | 1:100-1:500 |
Host / Isotype | Mouse / IgG2b |
Class | Monoclonal |
Type | Antibody |
Immunogen | Recombinant human SNF2H (SWI/SNF-related, matrix-associated, actin-dependent) protein. |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | 2 mg/mL |
Storage conditions | -20°C or -80°C if preferred |
RRID | AB_2610195 |
SMARCA5 is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. SMARCA5 is a component of the chromatin remodeling and spacing factor RSF, a facilitator of the transcription of class II genes by RNA polymerase II. The protein encoded by this gene is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The protein encoded by this gene is a component of the chromatin remodeling and spacing factor RSF, a facilitator of the transcription of class II genes by RNA polymerase II. The encoded protein is similar in sequence to the Drosophila ISWI chromatin remodeling protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.