Abnova
Manufacturer Code:MAB19675
Catalog # 89-336-199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | GFB-19 |
Immunogen | A synthetic peptide corresponding to human SMARCA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C for one year. After reconstitution at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: ATP-dependent helicase SMARCA4; BAF190 BRG1 FLJ39786 SNF2 SNF2-BETA SNF2L4 SNF2LB SWI2 hSNF2b; BAF190A; brahma protein-like 1; BRG1-associated factor 190A; BRM/SWI2-related gene 1; global transcription activator homologous sequence; homeotic gene regulator; Mitotic growth and transcription activator; nuclear protein GRB1; Protein brahma homolog 1; Protein BRG-1; SNF2-beta; SNF2-like 4; sucrose nonfermenting-like 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; Transcription activator BRG1
Gene Aliases: BAF190; BAF190A; BRG1; CSS4; hSNF2b; MRD16; RTPS2; SMARCA4; SNF2; SNF2B; SNF2L4; SNF2LB; SWI2
UniProt ID: (Human) P51532
Entrez Gene ID: (Human) 6597
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.